Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ANK2 SIRNA Assays ›
          Gene Symbol:
          Ank2
          Gene Name:
          ankyrin 2, brain
          Gene Aliases:
          AI835472, AW491075, Ank-2
          Chromosome Location:
          Chr.3: 126921607 - 127499870 on Build GRCm38
          UniGene ID:
          Mm.220242
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 287260

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ank2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001034168.1 NP_001029340.1 7 2710
          NM_178655.3 NP_848770.2 22 3013
          GenBank mRNA AK032060.1 - 22 3013
          AK083826.1 - 7 1269
          BC043123.1 - 7 2710
          BC059251.1 - 22 2964 6405358

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of heart rate
          cellular calcium ion homeostasis
          signal transduction
          protein localization
          positive regulation of gene expression
          regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
          regulation of cardiac muscle contraction by calcium ion signaling
          regulation of protein stability
          T-tubule organization
          protein localization to cell surface
          cellular protein localization
          protein localization to M-band
          protein localization to T-tubule
          regulation of apoptotic process
          positive regulation of potassium ion transport
          protein stabilization
          regulation of calcium ion transport
          positive regulation of calcium ion transport
          regulation of cardiac muscle contraction
          cardiac muscle contraction
          protein localization to endoplasmic reticulum
          protein localization to plasma membrane
          protein targeting to plasma membrane
          regulation of cardiac muscle cell contraction
          atrial cardiac muscle cell action potential
          SA node cell action potential
          regulation of cardiac muscle cell membrane potential
          atrial cardiac muscle cell to AV node cell communication
          SA node cell to atrial cardiac muscle cell communication
          regulation of heart rate by cardiac conduction
          regulation of AV node cell action potential
          regulation of SA node cell action potential
          regulation of atrial cardiac muscle cell action potential
          positive regulation of potassium ion transmembrane transporter activity
          regulation of calcium ion transmembrane transporter activity
          positive regulation of calcium ion transmembrane transporter activity
          regulation of cation channel activity
          positive regulation of cation channel activity
          protein binding
          potassium channel regulator activity
          protein binding, bridging
          ion channel binding
          ATPase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix