Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRPM3 SIRNA Assays ›
Gene Symbol:
Trpm3
Gene Name:
transient receptor potential cation channel, subfamily M, member 3
Gene Aliases:
6330504P12Rik, 9330180E14, AU018608, B930001P07Rik, LTRPC3, MLSN2
Chromosome Location:
Chr.19: 22138181 - 22989897 on Build GRCm38
UniGene ID:
Mm.131943
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
287275
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Trpm3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001035239.2
NP_001030316.1
5
727
NM_001035240.2
NP_001030317.1
5
727
NM_001035241.2
NP_001030318.1
5
727
NM_001035242.1
NP_001030319.1
5
727
NM_001035243.2
NP_001030320.1
5
727
NM_001035244.1
NP_001030321.1
5
775
NM_001035245.1
NP_001030322.1
5
775
NM_001035246.1
NP_001030323.1
5
775
NM_177341.4
NP_796315.2
5
794
GenBank mRNA
AJ544532.1
-
5
727
AJ544533.1
-
5
727
AJ544534.1
-
5
727
AJ544535.1
-
5
727
AJ544536.1
-
5
727
AK044454.1
-
5
796
AK044533.1
-
5
573
AK046908.1
-
5
608
AK051934.1
-
5
573
AK052064.1
-
5
581
AK053673.1
-
5
612
AK077331.1
-
5
607
AK082141.1
-
5
609
AK084402.1
-
5
580
AK142504.1
-
5
608
AK161909.1
-
5
608
AK163890.1
-
5
602
AK173218.1
-
5
798
BC141400.1
-
5
770
9056013
BC141412.1
-
5
770
9056025
HM064407.1
-
5
819
HM064408.1
-
5
819
HM064409.1
-
5
819
HM064410.1
-
5
819
HM064411.1
-
5
819
JF706722.1
-
5
790
JX644970.1
-
5
819
JX644971.1
-
5
819
JX644972.1
-
5
819
JX644973.1
-
5
819
JX644974.1
-
5
819
JX644975.1
-
5
819
JX644976.1
-
5
819
JX644977.1
-
5
819
JX644978.1
-
5
819
JX644979.1
-
5
819
JX644980.1
-
5
819
JX644981.1
-
5
819
JX644982.1
-
5
819
JX644983.1
-
5
819
JX644984.1
-
5
819
JX644985.1
-
5
819
JX644986.1
-
5
819
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cation transport
cation transmembrane transport
cation channel activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0