Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRPM3 SIRNA Assays ›
Gene Symbol:
Trpm3
Gene Name:
transient receptor potential cation channel, subfamily M, member 3
Gene Aliases:
6330504P12Rik, 9330180E14, AU018608, B930001P07Rik, LTRPC3, MLSN2
Chromosome Location:
Chr.19: 22138181 - 22989897 on Build GRCm38
UniGene ID:
Mm.131943
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
287276
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Trpm3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001035239.2
NP_001030316.1
4
506
NM_001035240.2
NP_001030317.1
4
506
NM_001035241.2
NP_001030318.1
4
506
NM_001035242.1
NP_001030319.1
4
506
NM_001035243.2
NP_001030320.1
4
506
NM_001035244.1
NP_001030321.1
4
554
NM_001035245.1
NP_001030322.1
4
554
NM_001035246.1
NP_001030323.1
4
554
NM_177341.4
NP_796315.2
4
573
GenBank mRNA
AJ544532.1
-
4
506
AJ544533.1
-
4
506
AJ544534.1
-
4
506
AJ544535.1
-
4
506
AJ544536.1
-
4
506
AK044454.1
-
4
575
AK044533.1
-
4
352
AK046908.1
-
4
387
AK051934.1
-
4
352
AK052064.1
-
4
360
AK053673.1
-
4
391
AK077331.1
-
4
386
AK082141.1
-
4
388
AK084402.1
-
4
359
AK085182.1
-
4
554
AK142504.1
-
4
387
AK161909.1
-
4
387
AK163890.1
-
4
381
AK173218.1
-
4
577
BC037449.1
-
4
402
BC141400.1
-
4
549
9056013
BC141412.1
-
4
549
9056025
HM064407.1
-
4
598
HM064408.1
-
4
598
HM064409.1
-
4
598
HM064410.1
-
4
598
HM064411.1
-
4
598
JF706722.1
-
4
569
JX644970.1
-
4
598
JX644971.1
-
4
598
JX644972.1
-
4
598
JX644973.1
-
4
598
JX644974.1
-
4
598
JX644975.1
-
4
598
JX644976.1
-
4
598
JX644977.1
-
4
598
JX644978.1
-
4
598
JX644979.1
-
4
598
JX644980.1
-
4
598
JX644981.1
-
4
598
JX644982.1
-
4
598
JX644983.1
-
4
598
JX644984.1
-
4
598
JX644985.1
-
4
598
JX644986.1
-
4
598
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001191562.
Gene Ontology Categories:
Function(s)
Process(es)
cation transport
cation transmembrane transport
cation channel activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-867f8cf58d-sfwzd:80/100.66.130.62:80.
git-commit: 7a7e1ef32845d1da800a461bcf18a29ea3e3afc0
git-url: https://github.com/thermofisher/magellan-search
git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix