Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MIER1 SIRNA Assays ›
Gene Symbol:
MIER1
Gene Name:
MIER1 transcriptional regulator
Gene Aliases:
ER1, MI-ER1
Chromosome Location:
Chr.1: 66924895 - 66988619 on Build GRCh38
UniGene ID:
Hs.605432
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
28912
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MIER1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077700.2
NP_001071168.2
3, 4
338
NM_001077701.2
NP_001071169.1
2, 3
194
NM_001077702.2
NP_001071170.2
4, 5
412
NM_001077703.2
NP_001071171.2
3, 4
338
NM_001077704.2
NP_001071172.1
2, 3
194
NM_001146110.1
NP_001139582.1
4, 5
415
NM_001146111.1
NP_001139583.1
4, 5
412
NM_001146112.1
NP_001139584.1
4, 5
373
NM_001278215.1
NP_001265144.1
4, 5
412
NM_020948.3
NP_065999.2
4, 5
412
XM_005271076.3
XP_005271133.1
4, 5
450
XM_011541864.2
XP_011540166.1
5, 6
357
XM_011541865.2
XP_011540167.1
5, 6
318
XM_017001919.1
XP_016857408.1
5, 6
334
XM_017001920.1
XP_016857409.1
5, 6
351
XM_017001921.1
XP_016857410.1
5, 6
379
XM_017001922.1
XP_016857411.1
5, 6
354
XM_017001924.1
XP_016857413.1
4, 5
306
XM_017001925.1
XP_016857414.1
4, 5
411
XM_017001926.1
XP_016857415.1
5, 6
350
XM_017001927.1
XP_016857416.1
3, 4
223
XM_017001928.1
XP_016857417.1
3, 4
299
XM_017001929.1
XP_016857418.1
4, 5
280
XM_017001930.1
XP_016857419.1
3, 4
224
XM_017001931.1
XP_016857420.1
4, 5
258
GenBank mRNA
AF515446.1
-
2, 3
194
AF515447.1
-
3, 4
275
AF515448.2
-
4, 5
349
AK302061.1
-
3, 4
338
AK307717.1
-
4, 5
309
AL831987.2
-
4, 5
354
AY124186.2
-
4, 5
349
AY124187.1
-
3, 4
275
AY124188.1
-
2, 3
194
AY124189.2
-
4, 5
349
AY124190.1
-
3, 4
275
AY124191.1
-
2, 3
194
AY124192.2
-
4, 5
349
AY124193.1
-
3, 4
275
AY124194.1
-
2, 3
194
BC017423.1
-
2, 3
171
BC108726.1
-
4, 5
293
5194296
CR627441.1
-
4, 5
249
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
signal transduction
positive regulation of chromatin silencing
positive regulation of I-kappaB kinase/NF-kappaB signaling
DNA binding
signal transducer activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0