Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRPM3 SIRNA Assays ›
Gene Symbol:
TRPM3
Gene Name:
transient receptor potential cation channel subfamily M member 3
Gene Aliases:
GON-2, LTRPC3, MLSN2
Chromosome Location:
Chr.9: 70529063 - 71446989 on Build GRCh38
UniGene ID:
Hs.47288
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
28914
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TRPM3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001007470.1
NP_001007471.1
4, 5
580
NM_001007471.2
NP_001007472.2
4, 5
928
NM_020952.4
NP_066003.3
4, 5
580
NM_024971.5
NP_079247.5
4, 5
580
NM_206944.3
NP_996827.3
4, 5
580
NM_206945.3
NP_996828.3
4, 5
580
NM_206946.3
NP_996829.3
4, 5
580
NM_206947.3
NP_996830.3
4, 5
580
NM_206948.2
NP_996831.1
4, 5
580
XM_011519035.2
XP_011517337.1
4, 5
805
XM_011519036.2
XP_011517338.1
4, 5
3730
XM_011519037.2
XP_011517339.1
4, 5
3716
XM_011519038.2
XP_011517340.1
4, 5
3713
XM_011519039.2
XP_011517341.1
4, 5
3714
XM_011519040.2
XP_011517342.1
4, 5
811
XM_011519041.2
XP_011517343.1
4, 5
3713
XM_011519042.2
XP_011517344.1
4, 5
811
XM_011519043.2
XP_011517345.1
4, 5
844
XM_011519044.2
XP_011517346.1
4, 5
3714
XM_011519045.2
XP_011517347.1
4, 5
805
XM_011519046.2
XP_011517348.1
4, 5
2508
XM_011519047.2
XP_011517349.1
4, 5
2478
XM_017015141.1
XP_016870630.1
4, 5
805
XM_017015142.1
XP_016870631.1
4, 5
805
XM_017015143.1
XP_016870632.1
4, 5
805
XM_017015144.1
XP_016870633.1
4, 5
571
XM_017015145.1
XP_016870634.1
4, 5
1249
XM_017015146.1
XP_016870635.1
4, 5
647
XM_017015147.1
XP_016870636.1
4, 5
2682
XM_017015148.1
XP_016870637.1
4, 5
805
XM_017015149.1
XP_016870638.1
4, 5
805
XM_017015150.1
XP_016870639.1
4, 5
805
XM_017015151.1
XP_016870640.1
4, 5
823
XM_017015152.1
XP_016870641.1
4, 5
2483
XM_017015153.1
XP_016870642.1
4, 5
815
XM_017015154.1
XP_016870643.1
4, 5
951
XM_017015155.1
XP_016870644.1
4, 5
2480
XM_017015157.1
XP_016870646.1
4, 5
2478
XM_017015158.1
XP_016870647.1
4, 5
2476
XM_017015159.1
XP_016870648.1
4, 5
571
XM_017015160.1
XP_016870649.1
4, 5
661
XM_017015161.1
XP_016870650.1
4, 5
805
GenBank mRNA
AB099661.1
-
1, 2
225
AB099662.1
-
1, 2
225
AB099663.1
-
1, 2
225
AB099664.1
-
1, 2
225
AB099665.1
-
1, 2
225
AF325212.1
-
4, 5
551
AF536748.1
-
1, 2
225
AF536749.1
-
1, 2
225
AF536750.1
-
1, 2
225
AF536751.1
-
1, 2
225
AF536752.1
-
1, 2
225
AF536753.1
-
1, 2
225
AJ505025.1
-
4, 5
928
AJ505026.1
-
4, 5
928
AK021788.1
-
4, 5
571
AK225732.1
-
4, 5
571
AK308682.1
-
4, 5
756
AL136545.1
-
4, 5
761
BC022454.2
-
3, 4
510
BC067733.1
-
4, 5
542
BC094699.1
-
4, 5
574
BC121821.1
-
2, 3
426
BC134414.1
-
2, 3
429
BC142972.1
-
4, 5
597
CD104154.1
-
4, 5
576
KF987075.1
-
4, 5
636
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cation transport
detection of temperature stimulus
sensory perception of temperature stimulus
protein tetramerization
calcium ion transmembrane transport
calcium activated cation channel activity
cation channel activity
calcium channel activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0