Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other KIR2DL4 SIRNA Assays ›
          Gene Symbol:
          KIR2DL4
          Gene Name:
          killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 4
          Gene Aliases:
          CD158D, G9P, KIR, KIR-103AS, KIR-2DL4, KIR103, KIR103AS
          Chromosome Location:
          Chr.19: 54803612 - 54814517 on Build GRCh38
          UniGene ID:
          Hs.661219
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 289349

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: KIR2DL4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001080770.1 NP_001074239.1 3, 4 398
          NM_001080772.1 NP_001074241.1 3, 4 398
          NM_002255.5 NP_002246.5 3, 4 398
          GenBank mRNA AF002979.1 - 3, 4 358
          AF002980.1 - 3, 4 370
          AF002981.1 - 3, 4 370
          AF002982.1 - 3, 4 370
          AF034771.1 - 3, 4 364
          AF034772.1 - 3, 4 364
          AF034773.1 - 3, 4 364
          AF276292.1 - 3, 4 358
          AF285436.1 - 3, 4 385
          AK309531.1 - 4, 5 634
          AY223512.2 - 1, 2 285
          AY223513.1 - 1, 2 285
          AY223514.1 - 1, 2 285
          AY223515.1 - 1, 2 285
          AY250088.1 - 1, 2 285
          AY359817.1 - 3, 4 369
          AY601812.1 - 3, 4 352
          AY789058.1 - 3, 4 352
          AY789059.1 - 3, 4 352
          AY789060.1 - 3, 4 352
          AY789061.1 - 3, 4 352
          AY789062.1 - 3, 4 352
          BC041611.1 - 3, 4 399 5485491
          BM918119.1 - 398
          DQ266438.1 - 3, 4 361
          EU194340.1 - 3, 4 358
          EU194342.1 - 3, 4 358
          U71199.1 - 3, 4 370
          U73394.1 - 3, 4 370
          X97229.1 - 3, 4 363
          X99479.1 - 3, 4 358
          X99480.1 - 3, 4 369
          Y13054.1 - 3, 4 358

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          cellular defense response
          signal transduction
          regulation of immune response
          transmembrane signaling receptor activity
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6bbccd986b-mx52t:80/100.66.130.62:80.
          git-commit: 717389b07c2b9f75290ff02e811d41bb86d4254a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0