Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LILRB1 SIRNA Assays ›
Gene Symbol:
LILRB1
Gene Name:
leukocyte immunoglobulin like receptor B1
Gene Aliases:
CD85J, ILT-2, ILT2, LIR-1, LIR1, MIR-7, MIR7, PIR-B, PIRB
Chromosome Location:
Chr.19: 54616930 - 54637925 on Build GRCh38
UniGene ID:
Hs.667388
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
289610
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LILRB1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001081637.2
NP_001075106.2
7
1295
NM_001081638.3
NP_001075107.2
7
1401
NM_001081639.3
NP_001075108.2
7
1401
NM_001278398.2
NP_001265327.2
7
1295
NM_001278399.2
NP_001265328.2
7
1295
NM_006669.6
NP_006660.4
8
1715
NR_103518.2
7
1295
XM_011526331.2
XP_011524633.1
6
1233
XM_011526332.2
XP_011524634.1
6
1233
XM_011526335.2
XP_011524637.1
6
1233
XM_011526336.2
XP_011524638.1
6
1203
XM_011526338.2
XP_011524640.1
7
1451
XM_017026182.1
XP_016881671.1
6
1230
XM_017026183.1
XP_016881672.1
6
1230
XM_017026184.1
XP_016881673.1
6
1233
XM_017026185.1
XP_016881674.1
6
1202
XM_017026186.1
XP_016881675.1
7
1355
XM_017026187.1
XP_016881676.1
8
1706
XM_017026188.1
XP_016881677.1
7
1355
XM_017026189.1
XP_016881678.1
7
1355
XM_017026190.1
XP_016881679.1
8
1710
XM_017026191.1
XP_016881680.1
6
1203
XM_017026192.1
XP_016881681.1
7
1415
GenBank mRNA
AF009005.1
-
6
1130
AF009006.1
-
6
1130
AF009007.1
-
6
1130
AF009220.1
-
6
1130
AF009221.1
-
6
1130
AF283984.1
-
7
1305
AF283985.1
-
7
1235
AK094834.1
-
1
45
AK223310.1
-
7
1328
AK292028.1
-
8
1425
AK292156.1
-
8
1715
BC015731.1
-
8
1470
4878915
DA991480.1
-
1
65
EU915608.1
-
6
1130
U82279.1
-
6
1130
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of T cell mediated cytotoxicity
positive regulation of defense response to virus by host
adaptive immune response
T cell proliferation involved in immune response
negative regulation of cytokine secretion involved in immune response
immune response-inhibiting cell surface receptor signaling pathway
Fc receptor mediated inhibitory signaling pathway
signal transduction
response to virus
positive regulation of gene expression
negative regulation of serotonin secretion
receptor internalization
interferon-gamma production
negative regulation of interferon-gamma production
negative regulation of mononuclear cell proliferation
negative regulation of interferon-beta secretion
negative regulation of T cell proliferation
negative regulation of tumor necrosis factor biosynthetic process
positive regulation of apoptotic process
negative regulation of interferon-gamma biosynthetic process
negative regulation of cell cycle
negative regulation of endocytosis
positive regulation of cytolysis
positive regulation of transcription from RNA polymerase II promoter
negative regulation of natural killer cell mediated cytotoxicity
negative regulation of alpha-beta T cell activation
regulation of immune response
defense response to virus
negative regulation of calcium ion transport
cellular response to lipopolysaccharide
interferon-gamma secretion
dendritic cell differentiation
negative regulation of dendritic cell apoptotic process
negative regulation of interleukin-10 secretion
negative regulation of interleukin-12 secretion
negative regulation of CD8-positive, alpha-beta T cell activation
negative regulation of T cell activation via T cell receptor contact with antigen bound to MHC molecule on antigen presenting cell
positive regulation of gamma-delta T cell activation involved in immune response
negative regulation of dendritic cell differentiation
negative regulation of transforming growth factor-beta secretion
negative regulation of osteoclast development
protein phosphatase 1 binding
HLA-A specific inhibitory MHC class I receptor activity
HLA-B specific inhibitory MHC class I receptor activity
MHC class I receptor activity
SH2 domain binding
MHC class I protein binding
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-2025.07.28-1.0