Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1468 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020854.3 | NP_065905.2 | 27, 28 | 3682 | ||
XM_005266730.2 | XP_005266787.1 | 27, 28 | 3729 | |||
XM_011526110.1 | XP_011524412.1 | 28, 29 | 3753 | |||
XM_011526111.1 | XP_011524413.1 | 28, 29 | 3753 | |||
XM_011526117.2 | XP_011524419.1 | 18, 19 | 5659 | |||
GenBank mRNA | AB040901.1 | - | 24, 25 | 2899 | ||
AK025228.1 | - | 2, 3 | 145 | |||
AK315400.1 | - | 23, 24 | 2626 | |||
BC011992.1 | - | 24, 25 | 2783 | 4339646 | ||
BX648481.1 | - | 27, 28 | 3665 |