Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BAIAP2 SIRNA Assays ›
          Gene Symbol:
          BAIAP2
          Gene Name:
          BAI1 associated protein 2
          Gene Aliases:
          BAP2, FLAF3, IRSP53
          Chromosome Location:
          Chr.17: 81035147 - 81117432 on Build GRCh38
          UniGene ID:
          Hs.128316
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 289781

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BAIAP2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001144888.1 NP_001138360.1 8 901
          NM_006340.2 NP_006331.1 8 901
          NM_017450.2 NP_059344.1 8 901
          NM_017451.2 NP_059345.1 8 901
          XM_005256943.1 XP_005257000.1 9 1000
          XM_005256944.1 XP_005257001.1 9 1000
          XM_005256945.1 XP_005257002.1 9 1000
          XM_005256948.3 XP_005257005.1 8 901
          XM_006721635.1 XP_006721698.1 9 1000
          XM_006721636.1 XP_006721699.1 9 1000
          XM_006721637.2 XP_006721700.1 8 901
          XM_011524193.1 XP_011522495.1 9 1000
          XM_011524194.1 XP_011522496.1 9 1000
          XM_011524195.1 XP_011522497.1 3 660
          XM_011524196.1 XP_011522498.1 3 422
          XM_011524197.1 XP_011522499.1 3 422
          XM_017024017.1 XP_016879506.1 7 1658
          XM_017024018.1 XP_016879507.1 6 1120
          XM_017024019.1 XP_016879508.1 3 914
          GenBank mRNA AB015019.1 - 8 886
          AB015020.1 - 8 886
          AB017119.1 - 8 886
          AB017120.1 - 8 886
          AK023688.1 - 9 986
          AK056888.1 - 4 1073
          AK222670.1 - 8 897
          AK225612.1 - 8 898
          AK297644.1 - 8 830
          AK298629.1 - 6 736
          AK301436.1 - 3 382
          BC002495.2 - 8 876
          BC014020.2 - 8 888 4125501
          BC032559.1 - 8 899 5562784
          U70669.1 - 8 886

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          plasma membrane organization
          axonogenesis
          insulin receptor signaling pathway
          regulation of cell shape
          response to bacterium
          dendrite development
          positive regulation of actin filament polymerization
          regulation of actin cytoskeleton organization
          Fc-gamma receptor signaling pathway involved in phagocytosis
          vascular endothelial growth factor receptor signaling pathway
          regulation of synaptic plasticity
          actin filament bundle assembly
          actin crosslink formation
          positive regulation of dendritic spine morphogenesis
          cell-cell adhesion
          positive regulation of actin cytoskeleton reorganization
          protein binding
          protein C-terminus binding
          cytoskeletal adaptor activity
          PDZ domain binding
          identical protein binding
          proline-rich region binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix