Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF250 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109689.3 | NP_001103159.1 | 3, 4 | 285 | ||
NM_021061.4 | NP_066405.1 | 3, 4 | 300 | |||
XM_005272327.1 | XP_005272384.1 | 3, 4 | 312 | |||
XM_005272328.3 | XP_005272385.1 | 3, 4 | 785 | |||
XM_006716612.3 | XP_006716675.1 | 3, 4 | 800 | |||
XM_006716613.3 | XP_006716676.1 | 3, 4 | 339 | |||
XM_011517209.1 | XP_011515511.1 | 3, 4 | 312 | |||
GenBank mRNA | AB208810.1 | - | 3, 4 | 255 | ||
AK054888.1 | - | 3, 4 | 300 | |||
AK095705.1 | - | 3, 4 | 243 | |||
AK294282.1 | - | 3, 4 | 261 | |||
BC017091.1 | - | 3, 4 | 282 | 3851128 | ||
BC107579.1 | - | 3, 4 | 289 | |||
DB206757.1 | - | 3, 4 | 285 | |||
DB272357.1 | - | 3, 4 | 261 | |||
DC314684.1 | - | 3, 4 | 261 |