Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MIER1 SIRNA Assays ›
Gene Symbol:
MIER1
Gene Name:
MIER1 transcriptional regulator
Gene Aliases:
ER1, MI-ER1
Chromosome Location:
Chr.1: 66924895 - 66988619 on Build GRCh38
UniGene ID:
Hs.605432
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
29001
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MIER1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077700.2
NP_001071168.2
9
1040
NM_001077701.2
NP_001071169.1
8
896
NM_001077702.2
NP_001071170.2
10
1114
NM_001077703.2
NP_001071171.2
9
1040
NM_001077704.2
NP_001071172.1
8
896
NM_001146110.1
NP_001139582.1
10
1117
NM_001146111.1
NP_001139583.1
10
1114
NM_001146112.1
NP_001139584.1
10
1075
NM_001146113.1
NP_001139585.1
8
894
NM_020948.3
NP_065999.2
10
1114
XM_005271076.3
XP_005271133.1
10
1152
XM_011541864.2
XP_011540166.1
11
1059
XM_011541865.2
XP_011540167.1
11
1020
XM_017001919.1
XP_016857408.1
11
1036
XM_017001920.1
XP_016857409.1
11
1053
XM_017001921.1
XP_016857410.1
11
1081
XM_017001922.1
XP_016857411.1
11
1056
XM_017001923.1
XP_016857412.1
9
876
XM_017001924.1
XP_016857413.1
10
1008
XM_017001925.1
XP_016857414.1
10
1113
XM_017001926.1
XP_016857415.1
11
1052
XM_017001927.1
XP_016857416.1
9
925
XM_017001928.1
XP_016857417.1
9
1001
XM_017001929.1
XP_016857418.1
10
982
XM_017001930.1
XP_016857419.1
9
926
XM_017001931.1
XP_016857420.1
10
960
XM_017001932.1
XP_016857421.1
8
889
XM_017001933.1
XP_016857422.1
7
743
XM_017001934.1
XP_016857423.1
6
639
GenBank mRNA
AB046830.1
-
6
665
AF515446.1
-
8
896
AF515447.1
-
9
977
AF515448.2
-
10
1051
AK302061.1
-
9
1040
AK307717.1
-
10
1011
AL831987.2
-
10
1056
AY124186.2
-
10
1051
AY124187.1
-
9
977
AY124188.1
-
8
896
AY124189.2
-
10
1051
AY124190.1
-
9
977
AY124191.1
-
8
896
AY124192.2
-
10
1051
AY124193.1
-
9
977
AY124194.1
-
8
896
BC108726.1
-
10
995
5194296
BC125217.1
-
7
692
40123797
BC125218.1
-
7
692
40123803
CR627441.1
-
10
951
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
signal transduction
positive regulation of chromatin silencing
positive regulation of I-kappaB kinase/NF-kappaB signaling
DNA binding
signal transducer activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-2025.07.28-1.0