Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF438 SIRNA Assays ›
Gene Symbol:
ZNF438
Gene Name:
zinc finger protein 438
Gene Aliases:
bA330O11.1
Chromosome Location:
Chr.10: 30844634 - 31032406 on Build GRCh38
UniGene ID:
Hs.660642
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
290251
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF438
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001143766.1
NP_001137238.1
7
989
NM_001143767.1
NP_001137239.1
5
802
NM_001143768.1
NP_001137240.1
6
860
NM_001143769.1
NP_001137241.1
7
1114
NM_001143770.1
NP_001137242.1
8
1040
NM_001143771.1
NP_001137243.1
7
1011
NM_182755.2
NP_877432.1
7
941
XM_006717393.3
XP_006717456.1
5
756
XM_006717396.3
XP_006717459.1
4
690
XM_006717398.3
XP_006717461.1
8
981
XM_006717399.3
XP_006717462.1
7
914
XM_011519371.2
XP_011517673.1
6
2964
XM_011519372.2
XP_011517674.1
6
991
XM_011519373.2
XP_011517675.1
5
2906
XM_011519374.2
XP_011517676.1
3
733
XM_011519376.2
XP_011517678.1
8
1066
XM_011519377.2
XP_011517679.1
8
1062
XM_017015863.1
XP_016871352.1
3
556
XM_017015864.1
XP_016871353.1
8
3200
XM_017015865.1
XP_016871354.1
7
3141
XM_017015866.1
XP_016871355.1
7
1031
XM_017015867.1
XP_016871356.1
6
2946
XM_017015868.1
XP_016871357.1
5
1252
XM_017015869.1
XP_016871358.1
5
2887
XM_017015870.1
XP_016871359.1
6
1786
XM_017015871.1
XP_016871360.1
5
1246
XM_017015872.1
XP_016871361.1
6
2944
XM_017015873.1
XP_016871362.1
7
3121
XM_017015874.1
XP_016871363.1
5
807
GenBank mRNA
AF428258.1
-
6
836
AK131357.1
-
7
1114
AK292730.1
-
7
907
AL833056.1
-
2
550
BC101622.1
-
3
566
8069128
BC104757.1
-
3
566
8143760
BC143462.1
-
4
665
BC143464.1
-
4
745
DQ356011.1
-
7
935
DQ356012.1
-
7
926
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0