Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEATR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022070.4 | NP_071353.4 | 6 | 746 | ||
XM_005257570.3 | XP_005257627.1 | 7 | 934 | |||
XM_011525110.2 | XP_011523412.1 | 5 | 633 | |||
XM_011525111.2 | XP_011523413.1 | 5 | 595 | |||
XM_017024936.1 | XP_016880425.1 | 6 | 1007 | |||
XM_017024937.1 | XP_016880426.1 | 6 | 2683 | |||
XM_017024938.1 | XP_016880427.1 | 6 | 736 | |||
XM_017024939.1 | XP_016880428.1 | 6 | 1007 | |||
GenBank mRNA | AF349752.1 | - | 7 | 691 | ||
AK127708.1 | - | 6 | 740 | |||
AK314655.1 | - | 6 | 736 | |||
BC119771.1 | - | 6 | 736 | 40108435 | ||
BC119813.1 | - | 6 | 737 | 40108426 | ||
BX640819.1 | - | 6 | 737 | |||
BX640831.1 | - | 6 | 737 | |||
CR627418.1 | - | 6 | 737 |