Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ICA1 SIRNA Assays ›
          Gene Symbol:
          ICA1
          Gene Name:
          islet cell autoantigen 1
          Gene Aliases:
          ICA69, ICAp69
          Chromosome Location:
          Chr.7: 8113184 - 8262687 on Build GRCh38
          UniGene ID:
          Hs.487561
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 29879

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ICA1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001136020.2 NP_001129492.1 3 380
          NM_001276478.1 NP_001263407.1 3 362
          NM_004968.3 NP_004959.2 3 365
          NM_022307.2 NP_071682.1 3 180
          XM_005249735.3 XP_005249792.1 3 302
          XM_011515345.1 XP_011513647.1 3 380
          XM_011515346.1 XP_011513648.1 3 365
          XM_011515347.2 XP_011513649.1 3 252
          XM_011515348.1 XP_011513650.1 3 298
          XM_011515349.1 XP_011513651.1 3 231
          XM_011515350.1 XP_011513652.1 3 302
          XM_011515351.1 XP_011513653.1 3 437
          XM_011515352.1 XP_011513654.1 3 256
          XM_011515353.2 XP_011513655.1 3 309
          XM_017012116.1 XP_016867605.1 3 148
          XM_017012117.1 XP_016867606.1 3 338
          XM_017012118.1 XP_016867607.1 3 298
          XM_017012119.1 XP_016867608.1 3 273
          XM_017012120.1 XP_016867609.1 3 319
          XM_017012121.1 XP_016867610.1 3 186
          XM_017012122.1 XP_016867611.1 3 351
          XM_017012123.1 XP_016867612.1 3 290
          XM_017012124.1 XP_016867613.1 3 451
          XM_017012125.1 XP_016867614.1 3 201
          GenBank mRNA AK292106.1 - 3 249
          BC008640.2 - 3 297 3854722
          DA076379.1 - 3 366
          DA154134.1 - 3 380
          DB010744.1 - 3 349
          DB040266.1 - 3 308
          EU416199.1 - 2 113
          L01100.1 - 3 291
          L21181.1 - 2 189
          U26591.1 - 3 180
          U26592.1 - 3 237
          U37183.1 - 1 79
          U38260.1 - 3 282

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          neurotransmitter transport
          regulation of protein homodimerization activity
          regulation of insulin secretion
          molecular_function
          protein binding
          protein domain specific binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix