Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other STRA6 SIRNA Assays ›
Gene Symbol:
STRA6
Gene Name:
stimulated by retinoic acid 6
Gene Aliases:
MCOPCB8, MCOPS9, PP14296
Chromosome Location:
Chr.15: 74179466 - 74212267 on Build GRCh38
UniGene ID:
Hs.24553
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
29904
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: STRA6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142617.1
NP_001136089.1
7, 8
842
NM_001142618.1
NP_001136090.1
7, 8
824
NM_001142619.1
NP_001136091.1
7, 8
815
NM_001199040.1
NP_001185969.1
7, 8
958
NM_001199041.1
NP_001185970.1
7, 8
969
NM_001199042.1
NP_001185971.1
7, 8
1073
NM_022369.3
NP_071764.3
7, 8
778
XM_011521883.1
XP_011520185.1
7, 8
744
XM_011521884.1
XP_011520186.1
7, 8
885
XM_011521885.2
XP_011520187.1
7, 8
719
XM_017022478.1
XP_016877967.1
7, 8
693
XM_017022479.1
XP_016877968.1
7, 8
784
XM_017022480.1
XP_016877969.1
7, 8
808
GenBank mRNA
AF352728.1
-
7, 8
644
AF352729.1
-
7, 8
754
AF370419.2
-
7, 8
750
AK022603.1
-
3, 4
328
AK056125.1
-
7, 8
839
AK091152.1
-
7, 8
868
AK291966.1
-
7, 8
714
AK298921.1
-
7, 8
713
AK298928.1
-
5, 6
382
AK299191.1
-
7, 8
958
AK302932.1
-
7, 8
969
AK309676.1
-
7, 8
1053
AK310878.1
-
7, 8
701
AY358748.1
-
7, 8
754
AY359089.1
-
7, 8
644
BC025256.1
-
7, 8
750
4892821
BX537413.1
-
7, 8
720
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
retinoid metabolic process
blood vessel development
kidney development
pulmonary valve morphogenesis
ventricular septum development
heart development
learning
feeding behavior
lung development
adrenal gland development
female genitalia development
vocal learning
ear development
nose morphogenesis
lung alveolus development
positive regulation of behavior
digestive tract morphogenesis
embryonic digestive tract development
developmental growth
smooth muscle tissue development
artery morphogenesis
cognition
neuromuscular process
vitamin transport
head development
head morphogenesis
face morphogenesis
lung vasculature development
diaphragm development
embryonic camera-type eye formation
eyelid development in camera-type eye
uterus morphogenesis
alveolar primary septum development
pulmonary artery morphogenesis
paramesonephric duct development
ductus arteriosus closure
receptor activity
vitamin transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0