Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MTHFSD SIRNA Assays ›
Gene Symbol:
MTHFSD
Gene Name:
methenyltetrahydrofolate synthetase domain containing
Gene Aliases:
-
Chromosome Location:
Chr.16: 86530176 - 86555235 on Build GRCh38
UniGene ID:
Hs.343627
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
30184
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MTHFSD
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001159377.1
NP_001152849.1
2, 3
166
NM_001159378.1
NP_001152850.1
2, 3
166
NM_001159379.1
NP_001152851.1
2, 3
163
NM_001159380.1
NP_001152852.1
2, 3
130
NM_022764.2
NP_073601.2
2, 3
163
NR_027489.1
2, 3
159
NR_027490.1
2, 3
166
XM_005256101.1
XP_005256158.1
2, 3
206
XM_005256105.1
XP_005256162.1
2, 3
212
XM_005256106.1
XP_005256163.1
2, 3
121
XM_011523280.2
XP_011521582.1
2, 3
206
XM_011523282.2
XP_011521584.1
2, 3
127
XM_011523283.2
XP_011521585.1
2, 3
127
XM_011523284.2
XP_011521586.1
2, 3
166
XM_011523285.1
XP_011521587.1
2, 3
127
XM_011523286.1
XP_011521588.1
2, 3
227
XM_011523287.1
XP_011521589.1
2, 3
131
XM_011523289.1
XP_011521591.1
2, 3
227
XM_017023573.1
XP_016879062.1
2, 3
120
GenBank mRNA
AK023060.1
-
2, 3
141
AK023955.1
-
2, 3
120
AK096836.1
-
2, 3
159
AK297726.1
-
2, 3
163
BC013911.2
-
1, 2
97
BC112057.1
-
2, 3
146
8327525
BC112059.1
-
2, 3
146
8327527
BC143702.1
-
2, 3
131
9052216
BC143704.1
-
2, 3
131
BC143705.1
-
2, 3
130
BC143707.1
-
2, 3
134
9052221
Back To Top
More Information
Gene Ontology Categories:
Process(es)
nucleotide binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0