Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GRAMD3 SIRNA Assays ›
Gene Symbol:
GRAMD3
Gene Name:
GRAM domain containing 3
Gene Aliases:
NS3TP2
Chromosome Location:
Chr.5: 126360096 - 126494377 on Build GRCh38
UniGene ID:
Hs.363558
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
30468
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GRAMD3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001146319.1
NP_001139791.1
2
397
NM_001146320.1
NP_001139792.1
2
546
NM_001146321.1
NP_001139793.1
2
546
NM_001146322.1
NP_001139794.1
2
208
NM_023927.2
NP_076416.2
2
546
XM_005272057.3
XP_005272114.1
2
284
XM_005272058.3
XP_005272115.1
2
284
XM_005272059.4
XP_005272116.1
2
631
XM_005272060.3
XP_005272117.1
2
371
XM_005272061.3
XP_005272118.1
2
366
XM_005272062.3
XP_005272119.1
2
332
XM_005272063.3
XP_005272120.1
2
481
XM_005272064.3
XP_005272121.1
2
348
XM_011543591.2
XP_011541893.1
2
1219
XM_011543592.2
XP_011541894.1
2
1215
XM_011543593.2
XP_011541895.1
2
371
XM_011543594.2
XP_011541896.1
2
143
XM_011543595.2
XP_011541897.1
2
250
XM_017009781.1
XP_016865270.1
2
106
XM_017009782.1
XP_016865271.1
2
348
XM_017009783.1
XP_016865272.1
2
114
XM_017009784.1
XP_016865273.1
2
302
XM_017009785.1
XP_016865274.1
2
355
GenBank mRNA
AK024966.1
-
2
314
AK225829.1
-
2
314
AK293307.1
-
2
251
AK294656.1
-
2
265
AK296267.1
-
2
251
AK298009.1
-
2
131
AK298349.1
-
2
219
AK300154.1
-
2
342
AK303857.1
-
2
148
AK316288.1
-
2
361
AY116970.1
-
2
116
BC008590.1
-
2
279
4148772
DC371647.1
-
2
546
DC413835.1
-
2
162
DC424558.1
-
2
397
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0