Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM65A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193522.1 | NP_001180451.1 | 13 | 1897 | ||
NM_001193523.1 | NP_001180452.1 | 13 | 2010 | |||
NM_001193524.1 | NP_001180453.1 | 13 | 1923 | |||
NM_024519.3 | NP_078795.2 | 13 | 1888 | |||
XM_011523321.1 | XP_011521623.1 | 13 | 2702 | |||
XM_011523322.1 | XP_011521624.1 | 13 | 1897 | |||
XM_011523324.2 | XP_011521626.1 | 13 | 1957 | |||
XM_011523325.1 | XP_011521627.1 | 13 | 1912 | |||
XM_011523326.1 | XP_011521628.1 | 13 | 2111 | |||
XM_017023663.1 | XP_016879152.1 | 13 | 1897 | |||
XM_017023664.1 | XP_016879153.1 | 12 | 1783 | |||
GenBank mRNA | AB067517.1 | - | 12 | 1746 | ||
AK023787.1 | - | 8 | 1380 | |||
AK097087.1 | - | 12 | 2138 | |||
AK127792.1 | - | 13 | 1897 | |||
AK293748.1 | - | 13 | 1920 | |||
AK295677.1 | - | 13 | 1854 | |||
AK302868.1 | - | 3 | 1047 | |||
AK307467.1 | - | 13 | 1841 | |||
AL834312.1 | - | 3 | 953 | |||
BC054512.1 | - | 3 | 930 | |||
BC098587.1 | - | 7 | 1325 |