Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC102B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001093729.1 | NP_001087198.1 | 5 | 1014 | ||
NM_024781.2 | NP_079057.2 | 3 | 908 | |||
XM_011526174.2 | XP_011524476.1 | 2 | 806 | |||
XM_011526175.2 | XP_011524477.1 | 2 | 806 | |||
XM_017025973.1 | XP_016881462.1 | 5 | 1047 | |||
XM_017025974.1 | XP_016881463.1 | 3 | 1050 | |||
XM_017025975.1 | XP_016881464.1 | 6 | 1098 | |||
XM_017025976.1 | XP_016881465.1 | 5 | 1047 | |||
XM_017025977.1 | XP_016881466.1 | 2 | 806 | |||
XM_017025978.1 | XP_016881467.1 | 4 | 957 | |||
XM_017025979.1 | XP_016881468.1 | 5 | 1046 | |||
XM_017025980.1 | XP_016881469.1 | 6 | 1097 | |||
GenBank mRNA | AK027247.1 | - | 1 | 197 | ||
AK057282.1 | - | 3 | 849 | |||
AK225597.1 | - | 1 | 197 | |||
AK308114.1 | - | 5 | 993 | |||
AL833863.1 | - | 2 | 226 | |||
BC056269.1 | - | 2 | 204 | |||
BC126448.1 | - | 1 | 186 | 8992164 | ||
BC126450.1 | - | 1 | 186 | 8992166 | ||
CR749520.1 | - | 3 | 855 |