Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144875.1 | NP_001138347.1 | 3 | 291 | ||
NM_001308235.1 | NP_001295164.1 | 3 | 483 | |||
NM_001308236.1 | NP_001295165.1 | 3 | 306 | |||
NM_024872.2 | NP_079148.2 | 3 | 262 | |||
XM_005265985.3 | XP_005266042.1 | 3 | 319 | |||
XM_005265987.1 | XP_005266044.1 | 3 | 319 | |||
XM_011534654.2 | XP_011532956.1 | 3 | 196 | |||
XM_011534655.2 | XP_011532957.1 | 3 | 372 | |||
XM_011534656.2 | XP_011532958.1 | 3 | 173 | |||
GenBank mRNA | AK026223.1 | - | 3 | 257 | ||
AL583057.3 | - | 3 | 437 | |||
BC004564.2 | - | 3 | 250 | 4054381 | ||
BC004867.2 | - | 1 | 358 | |||
BI489608.1 | - | 3 | 283 | |||
CF138017.1 | - | 3 | 414 | |||
DA674730.1 | - | 3 | 263 | |||
DA931137.1 | - | 3 | 373 |