Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PQLC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146343.1 | NP_001139815.1 | 4 | 951 | ||
NM_001146345.1 | NP_001139817.1 | 5 | 1168 | |||
NM_025078.4 | NP_079354.2 | 6 | 1222 | |||
XM_005266770.2 | XP_005266827.1 | 5 | 1017 | |||
XM_011526188.1 | XP_011524490.1 | 4 | 833 | |||
XM_017025997.1 | XP_016881486.1 | 7 | 1309 | |||
XM_017025998.1 | XP_016881487.1 | 6 | 1255 | |||
XM_017026001.1 | XP_016881490.1 | 5 | 1239 | |||
XM_017026003.1 | XP_016881492.1 | 6 | 1092 | |||
XM_017026004.1 | XP_016881493.1 | 5 | 1038 | |||
XM_017026005.1 | XP_016881494.1 | 5 | 957 | |||
GenBank mRNA | AK026031.1 | - | 3 | 798 | ||
AK123870.1 | - | 6 | 1093 | |||
AK126188.1 | - | 5 | 1793 | |||
AK301881.1 | - | 4 | 951 | |||
AK303818.1 | - | 3 | 733 | |||
BC030140.2 | - | 6 | 1186 | 4761671 |