Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025145.5 | NP_079421.5 | 6 | 943 | ||
XM_005270171.2 | XP_005270228.1 | 6 | 944 | |||
XM_005270172.3 | XP_005270229.1 | 6 | 944 | |||
XM_011540196.2 | XP_011538498.1 | 6 | 944 | |||
XM_011540197.2 | XP_011538499.1 | 6 | 944 | |||
XM_011540198.2 | XP_011538500.1 | 6 | 944 | |||
XM_011540199.2 | XP_011538501.1 | 6 | 944 | |||
XM_011540200.2 | XP_011538502.1 | 6 | 944 | |||
XM_011540201.2 | XP_011538503.1 | 6 | 944 | |||
XM_011540202.2 | XP_011538504.1 | 2 | 260 | |||
XM_011540203.2 | XP_011538505.1 | 4 | 486 | |||
XM_017016681.1 | XP_016872170.1 | 6 | 944 | |||
XM_017016682.1 | XP_016872171.1 | 6 | 944 | |||
XM_017016683.1 | XP_016872172.1 | 4 | 486 | |||
XM_017016684.1 | XP_016872173.1 | 6 | 944 | |||
GenBank mRNA | AK026597.1 | - | 6 | 921 | ||
AK295018.1 | - | 6 | 922 | |||
AL136901.1 | - | 6 | 885 | |||
BC036647.2 | - | 6 | 943 | 4825754 |