Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146102.1 | NP_001139574.1 | 8 | 1824 | ||
NM_001146103.1 | NP_001139575.1 | 8 | 1901 | |||
NM_001146104.1 | NP_001139576.1 | 8 | 1900 | |||
NM_001146105.1 | NP_001139577.1 | 8 | 1719 | |||
NM_001146106.1 | NP_001139578.1 | 8 | 1900 | |||
NM_031458.2 | NP_113646.2 | 8 | 2006 | |||
XM_005247820.2 | XP_005247877.1 | 8 | 1930 | |||
XM_011513218.1 | XP_011511520.1 | 8 | 1796 | |||
XM_017007304.1 | XP_016862793.1 | 8 | 1814 | |||
XM_017007305.1 | XP_016862794.1 | 8 | 1709 | |||
XM_017007306.1 | XP_016862795.1 | 8 | 1790 | |||
GenBank mRNA | AB209742.1 | - | 4 | 523 | ||
AF307338.1 | - | 8 | 1907 | |||
AF307339.1 | - | 8 | 1802 | |||
AK123412.1 | - | 2 | 762 | |||
AK130147.1 | - | 3 | 355 | |||
AK292959.1 | - | 8 | 1679 | |||
AK299107.1 | - | 8 | 1793 | |||
AK313494.1 | - | 8 | 1793 | |||
AL713679.1 | - | 1 | 107 | |||
AL832929.1 | - | 1 | 39 | |||
BC017463.1 | - | 8 | 1868 | |||
BC039580.1 | - | 7 | 1677 | 5416494 | ||
BP259272.1 | - | 3 | 278 | |||
BX648869.1 | - | 6 | 546 |