Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ZNF41 SIRNA Assays ›
          Gene Symbol:
          ZNF41
          Gene Name:
          zinc finger protein 41
          Gene Aliases:
          MRX89
          Chromosome Location:
          Chr.X: 47444691 - 47483671 on Build GRCh38
          UniGene ID:
          Hs.496074
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 3407

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ZNF41
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001324139.1 NP_001311068.1 3, 4 377
          NM_001324140.1 NP_001311069.1 4, 5 667
          NM_001324141.1 NP_001311070.1 3, 4 406
          NM_001324142.1 NP_001311071.1 4, 5 683
          NM_001324143.1 NP_001311072.1 3, 4 719
          NM_001324144.1 NP_001311073.1 4, 5 757
          NM_001324145.1 NP_001311074.1 3, 4 422
          NM_001324147.1 NP_001311076.1 4, 5 728
          NM_001324148.1 NP_001311077.1 4, 5 773
          NM_001324149.1 NP_001311078.1 3, 4 446
          NM_001324150.1 NP_001311079.1 5, 6 855
          NM_001324151.1 NP_001311080.1 4, 5 797
          NM_001324152.1 NP_001311081.1 4, 5 843
          NM_001324153.1 NP_001311082.1 4, 5 707
          NM_001324154.1 NP_001311083.1 3, 4 594
          NM_001324155.1 NP_001311084.1 3, 4 618
          NM_007130.3 NP_009061.1 4, 5 1070
          NM_153380.3 NP_700359.1 4, 5 943
          XM_006724550.3 XP_006724613.1 4, 5 796
          XM_006724555.3 XP_006724618.1 4, 5 1091
          XM_017029810.1 XP_016885299.1 4, 5 1171
          XM_017029811.1 XP_016885300.1 4, 5 1232
          XM_017029812.1 XP_016885301.1 4, 5 1153
          XM_017029813.1 XP_016885302.1 4, 5 702
          XM_017029814.1 XP_016885303.1 4, 5 1142
          XM_017029815.1 XP_016885304.1 4, 5 1191
          XM_017029816.1 XP_016885305.1 4, 5 630
          XM_017029817.1 XP_016885306.1 4, 5 590
          GenBank mRNA AJ010017.1 - 3, 4 386
          AJ010018.1 - 3, 4 410
          AJ010019.1 - 3, 4 456
          AJ010020.1 - 3, 4 472
          AJ010021.1 - 3, 4 496
          AK290021.1 - 4, 5 724
          AK294858.1 - 3, 4 319
          BC015023.1 - 4, 5 953 3908304
          BQ053634.1 - 4, 5 611
          DA567652.1 - 3, 4 316
          DA783869.1 - 3, 4 377
          X60155.2 - 4, 5 709

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-849d5b7584-dvwjr:80/100.66.130.62:80.
          git-commit: 0a9b6045e9144c2c4a21b698be73aa131cdbda4b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0