Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IFT122 SIRNA Assays ›
Gene Symbol:
IFT122
Gene Name:
intraflagellar transport 122
Gene Aliases:
CED, CED1, SPG, WDR10, WDR10p, WDR140
Chromosome Location:
Chr.3: 129440036 - 129520507 on Build GRCh38
UniGene ID:
Hs.655284
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
35654
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IFT122
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001280541.1
NP_001267470.1
3
453
NM_001280545.1
NP_001267474.1
3
453
NM_001280546.1
NP_001267475.1
3
453
NM_018262.3
NP_060732.2
3
453
NM_052985.3
NP_443711.2
3
453
NM_052989.2
NP_443715.1
3
453
NM_052990.2
NP_443716.1
3
453
XM_005247609.1
XP_005247666.1
3
418
XM_006713689.1
XP_006713752.1
3
418
XM_006713691.2
XP_006713754.1
3
418
XM_006713692.2
XP_006713755.1
3
418
XM_006713695.2
XP_006713758.1
3
418
XM_011512972.2
XP_011511274.1
3
418
XM_017006817.1
XP_016862306.1
3
418
XM_017006818.1
XP_016862307.1
3
418
XM_017006819.1
XP_016862308.1
3
418
XM_017006820.1
XP_016862309.1
3
418
XM_017006821.1
XP_016862310.1
3
418
XM_017006822.1
XP_016862311.1
3
365
XM_017006823.1
XP_016862312.1
3
418
XM_017006824.1
XP_016862313.1
3
418
XM_017006825.1
XP_016862314.1
3
418
XM_017006826.1
XP_016862315.1
3
418
XM_017006827.1
XP_016862316.1
3
418
XM_017006828.1
XP_016862317.1
3
365
XM_017006829.1
XP_016862318.1
3
365
XM_017006830.1
XP_016862319.1
3
265
XM_017006831.1
XP_016862320.1
3
414
XM_017006832.1
XP_016862321.1
3
242
XM_017006833.1
XP_016862322.1
3
365
XM_017006834.1
XP_016862323.1
3
365
XM_017006835.1
XP_016862324.1
3
414
XM_017006836.1
XP_016862325.1
3
265
GenBank mRNA
AF244930.1
-
3
350
AF244931.1
-
3
350
AF302154.1
-
3
235
AK001759.1
-
3
266
AK022525.1
-
3
364
AK023000.1
-
3
249
AK024435.1
-
2
141
AK124140.1
-
3
256
AK223095.1
-
3
265
AK293852.1
-
3
252
AK298526.1
-
3
177
AK299903.1
-
3
166
AK315103.1
-
3
277
AL133565.1
-
1
30
AU123134.1
-
3
364
BC003045.1
-
3
245
2823536
BC004238.2
-
3
245
2823536
BC028353.2
-
3
193
4836844
DB099554.1
-
3
453
DC343174.1
-
3
200
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_001057925, XM_006237067, BC087667.
Gene Ontology Categories:
Function(s)
Process(es)
neural tube closure
signal transduction downstream of smoothened
embryonic body morphogenesis
negative regulation of smoothened signaling pathway involved in ventral spinal cord patterning
embryonic heart tube development
embryonic forelimb morphogenesis
intraciliary anterograde transport
intraciliary retrograde transport
intraciliary transport involved in cilium morphogenesis
cilium assembly
embryonic digit morphogenesis
negative regulation of smoothened signaling pathway
camera-type eye morphogenesis
negative regulation of epithelial cell proliferation
limb development
cilium morphogenesis
ciliary receptor clustering involved in smoothened signaling pathway
smoothened signaling pathway involved in dorsal/ventral neural tube patterning
embryonic heart tube left/right pattern formation
protein localization to cilium
establishment of protein localization to organelle
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0