Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DPP8 SIRNA Assays ›
          Gene Symbol:
          DPP8
          Gene Name:
          dipeptidyl peptidase 8
          Gene Aliases:
          DP8, DPRP-1, DPRP1, MST097, MSTP097, MSTP135, MSTP141
          Chromosome Location:
          Chr.15: 65442463 - 65517713 on Build GRCh38
          UniGene ID:
          Hs.458609
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 36382

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DPP8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001320875.1 NP_001307804.1 9, 10 1399
          NM_001320876.1 NP_001307805.1 8, 9 1368
          NM_017743.5 NP_060213.2 8, 9 1263
          NM_130434.4 NP_569118.1 8, 9 1263
          NM_197960.3 NP_932064.1 9, 10 1304
          NM_197961.3 NP_932065.1 9, 10 1399
          NR_135485.1 8, 9 1263
          NR_135486.1 9, 10 1304
          XM_011521730.1 XP_011520032.1 7, 8 2523
          XM_011521731.1 XP_011520033.1 7, 8 2478
          XM_011521732.2 XP_011520034.1 8, 9 2652
          XM_011521733.1 XP_011520035.1 7, 8 1070
          XM_011521734.1 XP_011520036.1 8, 9 2652
          XM_017022373.1 XP_016877862.1 8, 9 2652
          XM_017022374.1 XP_016877863.1 8, 9 2652
          XM_017022375.1 XP_016877864.1 7, 8 1115
          XM_017022376.1 XP_016877865.1 8, 9 1244
          XM_017022377.1 XP_016877866.1 6, 7 941
          XM_017022378.1 XP_016877867.1 8, 9 2652
          XM_017022379.1 XP_016877868.1 7, 8 1070
          XM_017022380.1 XP_016877869.1 7, 8 1070
          XM_017022381.1 XP_016877870.1 6, 7 941
          GenBank mRNA AF221634.1 - 8, 9 1237
          AF221636.1 - 1, 2 74
          AK000290.1 - 8, 9 1186
          AK027826.1 - 4, 5 442
          AK291057.1 - 9, 10 1374
          AK296915.1 - 8, 9 1339
          AK302638.1 - 9, 10 1240
          AY172659.1 - 7, 8 1031
          AY354202.1 - 9, 10 1232
          BC030688.2 - 9, 10 1390 4822550

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          immune response
          serine-type peptidase activity
          dipeptidyl-peptidase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0