Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: M1AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281296.1 | NP_001268225.1 | 10 | 2107 | ||
NM_001321739.1 | NP_001308668.1 | 11 | 2119 | |||
NM_138804.4 | NP_620159.2 | 11 | 2110 | |||
XM_005264152.3 | XP_005264209.1 | 6 | 1267 | |||
XM_006711946.3 | XP_006712009.1 | 11 | 2455 | |||
XM_011532548.2 | XP_011530850.1 | 10 | 2444 | |||
XM_011532549.2 | XP_011530851.1 | 11 | 2397 | |||
XM_011532550.2 | XP_011530852.1 | 11 | 2066 | |||
XM_011532551.2 | XP_011530853.1 | 11 | 2065 | |||
XM_011532552.2 | XP_011530854.1 | 10 | 2282 | |||
XM_011532553.2 | XP_011530855.1 | 10 | 2250 | |||
XM_011532554.2 | XP_011530856.1 | 7 | 1279 | |||
GenBank mRNA | AK126374.1 | - | 6 | 1268 | ||
AK130161.1 | - | 6 | 1083 | |||
BC025997.1 | - | 11 | 2113 | 4823281 |