Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF627 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290083.1 | NP_001277012.1 | 5 | 597 | ||
NM_001290084.1 | NP_001277013.1 | 4 | 431 | |||
NM_001290085.1 | NP_001277014.1 | 4 | 438 | |||
NM_145295.3 | NP_660338.1 | 4 | 442 | |||
XM_011527780.2 | XP_011526082.1 | 4 | 330 | |||
GenBank mRNA | AK074846.1 | - | 4 | 350 | ||
AK095639.1 | - | 4 | 368 | |||
BC006279.2 | - | 4 | 363 | |||
BC069232.1 | - | 4 | 327 | 6645413 | ||
BC098416.1 | - | 4 | 413 | |||
DA786363.1 | - | 4 | 398 | |||
DB043358.1 | - | 4 | 442 | |||
DB158202.1 | - | 5 | 539 |