Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHLDB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134437.1 | NP_001127909.1 | 3 | 628 | ||
NM_001134438.1 | NP_001127910.1 | 2 | 686 | |||
NM_001134439.1 | NP_001127911.1 | 2 | 447 | |||
NM_145753.2 | NP_665696.1 | 2 | 686 | |||
GenBank mRNA | AB209903.1 | - | 2 | 328 | ||
AF506820.1 | - | 1 | 390 | |||
AJ496194.1 | - | 1 | 301 | |||
AK092996.1 | - | 2 | 372 | |||
AK096151.1 | - | 2 | 628 | |||
AK296841.1 | - | 3 | 628 | |||
AK307491.1 | - | 3 | 590 | |||
AK315050.1 | - | 2 | 598 | |||
AL832205.1 | - | 2 | 409 | |||
AL832476.1 | - | 2 | 375 | |||
BC038806.1 | - | 2 | 418 | |||
BC069194.1 | - | 2 | 575 | |||
BC142678.1 | - | 1 | 293 | 40148225 | ||
BC150210.1 | - | 1 | 293 | 40148898 | ||
CR749654.1 | - | 3 | 611 | |||
DA272187.1 | - | 2 | 447 |