Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM122B SIRNA Assays ›
Gene Symbol:
FAM122B
Gene Name:
family with sequence similarity 122B
Gene Aliases:
SPACIA2
Chromosome Location:
Chr.X: 134769566 - 134797232 on Build GRCh38
UniGene ID:
Hs.404706
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
38170
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM122B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001166599.2
NP_001160071.1
3
1142
NM_001166600.2
NP_001160072.1
5
511
NM_001170756.1
NP_001164227.1
4
399
NM_001170757.1
NP_001164228.1
4
227
NM_145284.5
NP_660327.2
3
342
XM_006724730.2
XP_006724793.1
4
1255
XM_006724732.2
XP_006724795.1
4
1255
XM_006724733.3
XP_006724796.1
3
1192
XM_011531282.1
XP_011529584.1
4
1267
XM_011531283.1
XP_011529585.1
4
1267
XM_011531284.1
XP_011529586.1
4
1255
XM_011531285.1
XP_011529587.1
4
1255
XM_011531286.1
XP_011529588.1
3
1210
XM_011531287.1
XP_011529589.1
4
1267
XM_011531288.1
XP_011529590.1
3
1192
XM_011531289.1
XP_011529591.1
4
1267
XM_011531290.1
XP_011529592.1
3
1198
XM_011531291.1
XP_011529593.1
4
1267
XM_011531292.1
XP_011529594.1
4
1267
XM_011531293.1
XP_011529595.1
4
1255
XM_011531294.1
XP_011529596.1
3
1198
XM_017029306.1
XP_016884795.1
3
1210
XM_017029307.1
XP_016884796.1
3
1210
GenBank mRNA
AB541014.1
-
4
249
AB541015.1
-
4
249
AK124650.1
-
5
484
AK124940.1
-
3
128
AK125991.1
-
4
302
AK292517.1
-
3
261
AK297720.1
-
4
227
AL832218.1
-
3
275
BC019221.2
-
3
1121
3841054
BC110846.1
-
3
1178
6046376
BX538218.1
-
4
619
CN303065.1
-
5
504
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-2025.07.28-1.0