Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF560 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152476.2 | NP_689689.2 | 5, 6 | 444 | ||
XM_011527696.2 | XP_011525998.1 | 5, 6 | 473 | |||
XM_011527697.2 | XP_011525999.1 | 5, 6 | 499 | |||
XM_017026327.1 | XP_016881816.1 | 5, 6 | 453 | |||
XM_017026328.1 | XP_016881817.1 | 5, 6 | 453 | |||
XM_017026329.1 | XP_016881818.1 | 5, 6 | 453 | |||
XM_017026330.1 | XP_016881819.1 | 5, 6 | 400 | |||
GenBank mRNA | AK056548.1 | - | 5, 6 | 444 | ||
BC101040.1 | - | 5, 6 | 409 | 40007878 | ||
BC101041.1 | - | 5, 6 | 409 | 40007880 | ||
BC101042.2 | - | 5, 6 | 409 | 40007881 | ||
DA744260.1 | - | 5, 6 | 444 |