Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC8P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003950.1 | 6 | 1454 | |||
GenBank mRNA | AK056130.1 | - | 6 | 1387 | ||
AK092530.1 | - | 5 | 1289 | |||
AK092581.1 | - | 3 | 1045 | |||
BC105936.1 | - | 2 | 797 | |||
BC105937.1 | - | 2 | 797 | |||
BC107600.1 | - | 2 | 797 |