Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145195.1 | NP_001138667.1 | 8 | 1608 | ||
NM_001282733.1 | NP_001269662.1 | 8 | 1608 | |||
NM_001282734.1 | NP_001269663.1 | 7 | 1261 | |||
NM_152725.3 | NP_689938.2 | 8 | 1608 | |||
GenBank mRNA | AK055061.1 | - | 8 | 1556 | ||
AK295294.1 | - | 7 | 1137 | |||
AK303735.1 | - | 8 | 1293 | |||
BC035118.1 | - | 8 | 1536 | |||
BC047635.1 | - | 8 | 1393 | 4820920 | ||
BC065917.1 | - | 7 | 1352 | |||
BC094700.1 | - | 8 | 1484 | 30407123 | ||
BC117323.1 | - | 7 | 1358 | 40125874 | ||
BC143551.1 | - | 7 | 1357 | 9052062 |