Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYCP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177948.1 | NP_001171419.1 | 2, 3 | 249 | ||
NM_001177949.1 | NP_001171420.1 | 2, 3 | 264 | |||
NM_153694.4 | NP_710161.1 | 2, 3 | 260 | |||
XM_005268922.4 | XP_005268979.3 | 2, 3 | 510 | |||
XM_005268924.1 | XP_005268981.1 | 2, 3 | 248 | |||
XM_005268925.1 | XP_005268982.1 | 2, 3 | 253 | |||
XM_005268926.3 | XP_005268983.1 | 2, 3 | 232 | |||
XM_005268927.2 | XP_005268984.1 | 2, 3 | 221 | |||
XM_011538421.2 | XP_011536723.2 | 2, 3 | 432 | |||
XM_017019368.1 | XP_016874857.1 | 2, 3 | 507 | |||
GenBank mRNA | AF492003.1 | - | 2, 3 | 205 | ||
AF517774.1 | - | 2, 3 | 203 | |||
BC062662.1 | - | 2, 3 | 255 | 6616222 | ||
BQ428625.1 | - | 2, 3 | 265 |