Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYSND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173555.3 | NP_775826.2 | 3, 4 | 1633 | ||
NR_073581.1 | 3, 4 | 521 | ||||
NR_073582.1 | 3, 4 | 401 | ||||
NR_073591.1 | 3, 4 | 312 | ||||
NR_073595.1 | 3, 4 | 557 | ||||
GenBank mRNA | AK074968.1 | - | 3, 4 | 459 | ||
AK097697.1 | - | 2, 3 | 1176 | |||
BC016840.1 | - | 3, 4 | 1087 | |||
BC111501.1 | - | 3, 4 | 1572 | 5732356 | ||
BI829894.1 | - | 3, 4 | 309 | |||
CV805418.1 | - | 3, 4 | 557 | |||
DA434127.1 | - | 3, 4 | 350 |