Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM129C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098524.1 | NP_001091994.1 | 14, 15 | 1894 | ||
NM_001321826.1 | NP_001308755.1 | 13, 14 | 1766 | |||
NM_001321827.1 | NP_001308756.1 | 13, 14 | 1766 | |||
NM_001321828.1 | NP_001308757.1 | 12, 13 | 1604 | |||
NM_173544.4 | NP_775815.2 | 14, 15 | 1894 | |||
XM_005259814.3 | XP_005259871.1 | 12, 13 | 1633 | |||
XM_011527781.2 | XP_011526083.1 | 13, 14 | 1741 | |||
XM_011527786.2 | XP_011526088.1 | 13, 14 | 1741 | |||
XM_017026453.1 | XP_016881942.1 | 13, 14 | 1741 | |||
XM_017026454.1 | XP_016881943.1 | 12, 13 | 1633 | |||
XM_017026455.1 | XP_016881944.1 | 12, 13 | 1613 | |||
XM_017026456.1 | XP_016881945.1 | 12, 13 | 1668 | |||
XM_017026457.1 | XP_016881946.1 | 10, 11 | 1318 | |||
GenBank mRNA | AK097121.1 | - | 13, 14 | 1741 | ||
AK298062.1 | - | 12, 13 | 1595 | |||
AK300046.1 | - | 13, 14 | 1748 | |||
AY254197.1 | - | 14, 15 | 1894 | |||
AY254198.1 | - | 14, 15 | 1894 | |||
AY254199.1 | - | 14, 15 | 1894 | |||
BC053606.1 | - | 12, 13 | 1592 |