Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ADGRB2 SIRNA Assays ›
Gene Symbol:
ADGRB2
Gene Name:
adhesion G protein-coupled receptor B2
Gene Aliases:
BAI2
Chromosome Location:
Chr.1: 31727105 - 31764063 on Build GRCh38
UniGene ID:
Hs.524138
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
4162
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ADGRB2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001294335.1
NP_001281264.1
19, 20
3254
NM_001294336.1
NP_001281265.1
19, 20
3254
XM_011541848.2
XP_011540150.1
18, 19
3208
XM_011541849.2
XP_011540151.1
17, 18
3043
XM_011541858.2
XP_011540160.1
16, 17
2207
XM_017001899.1
XP_016857388.1
18, 19
3338
XM_017001900.1
XP_016857389.1
17, 18
3310
XM_017001901.1
XP_016857390.1
17, 18
3310
XM_017001902.1
XP_016857391.1
17, 18
3311
XM_017001903.1
XP_016857392.1
16, 17
3242
XM_017001904.1
XP_016857393.1
17, 18
3307
XM_017001905.1
XP_016857394.1
17, 18
3309
XM_017001906.1
XP_016857395.1
17, 18
3308
XM_017001907.1
XP_016857396.1
16, 17
3142
XM_017001908.1
XP_016857397.1
16, 17
3143
XM_017001909.1
XP_016857398.1
16, 17
3144
XM_017001910.1
XP_016857399.1
16, 17
3142
XM_017001911.1
XP_016857400.1
16, 17
3143
XM_017001912.1
XP_016857401.1
15, 16
2976
GenBank mRNA
AB005298.1
-
19, 20
3226
AK296502.1
-
14, 15
1886
BC136533.1
-
17, 18
2922
9020523
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
peripheral nervous system development
negative regulation of angiogenesis
positive regulation of synapse assembly
G-protein coupled receptor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0