Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF226 SIRNA Assays ›
Gene Symbol:
ZNF226
Gene Name:
zinc finger protein 226
Gene Aliases:
-
Chromosome Location:
Chr.19: 44165062 - 44177685 on Build GRCh38
UniGene ID:
Hs.145956
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
42507
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF226
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001032372.1
NP_001027544.1
5
380
NM_001032373.1
NP_001027545.1
5
344
NM_001032374.1
NP_001027546.1
5
513
NM_001146220.2
NP_001139692.1
6
404
NM_001319088.1
NP_001306017.1
5
547
NM_001319089.1
NP_001306018.1
6
404
NM_001319090.1
NP_001306019.1
5
424
NM_015919.3
NP_057003.2
5
344
NM_016444.2
NP_057528.2
6
567
XM_005259227.2
XP_005259284.1
6
678
XM_006723367.3
XP_006723430.1
7
574
XM_006723368.3
XP_006723431.1
7
460
XM_006723369.2
XP_006723432.1
6
517
XM_017027262.1
XP_016882751.1
6
526
XM_017027263.1
XP_016882752.1
5
348
XM_017027264.1
XP_016882753.1
5
505
XM_017027265.1
XP_016882754.1
5
412
GenBank mRNA
AF070666.1
-
5
339
AF228418.1
-
6
565
AK023091.1
-
5
340
AK314724.1
-
6
363
BC017786.1
-
5
336
BC024197.2
-
5
380
3532273
BI457780.1
-
5
345
BX091242.1
-
5
506
BX537487.1
-
5
336
BX648775.1
-
5
508
CD700800.1
-
5
370
DA080156.1
-
5
508
DA178589.1
-
5
367
DA435005.1
-
6
350
DA558473.1
-
5
352
DB015503.1
-
5
533
DR002748.1
-
5
386
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0