Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAXO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287049.1 | NP_001273978.1 | 2 | 203 | ||
NM_001287050.1 | NP_001273979.1 | 2 | 445 | |||
NM_153707.3 | NP_714918.2 | 2 | 445 | |||
XM_011517744.1 | XP_011516046.1 | 3 | 525 | |||
XM_011517745.1 | XP_011516047.1 | 4 | 460 | |||
XM_011517746.2 | XP_011516048.1 | 4 | 381 | |||
XM_017014314.1 | XP_016869803.1 | 4 | 771 | |||
XM_017014315.1 | XP_016869804.1 | 2 | 158 | |||
GenBank mRNA | AK057474.1 | - | 2 | 440 | ||
AK292245.1 | - | 2 | 396 | |||
BC033489.1 | - | 2 | 373 | 5170510 | ||
BM563230.1 | - | 2 | 222 | |||
DB035807.1 | - | 2 | 439 | |||
DB038218.1 | - | 2 | 446 |