Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf198 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136494.1 | NP_001129966.1 | 3 | 236 | ||
NM_032800.2 | NP_116189.1 | 1 | 178 | |||
GenBank mRNA | AK027431.1 | - | 1 | 153 | ||
AK096166.1 | - | 3 | 236 | |||
BC066649.1 | - | 1 | 166 | 6011914 | ||
DA754292.1 | - | 1 | 178 |