Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXXC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317199.1 | NP_001304128.1 | 3 | 1471 | ||
NM_001317200.1 | NP_001304129.1 | 2 | 908 | |||
NM_001317201.1 | NP_001304130.1 | 2 | 960 | |||
NM_001317202.1 | NP_001304131.1 | 2 | 924 | |||
NM_001317203.1 | NP_001304132.1 | 3 | 1132 | |||
NM_001317204.1 | NP_001304133.1 | 3 | 1002 | |||
NM_001317205.1 | NP_001304134.1 | 2 | 1001 | |||
NM_001317206.1 | NP_001304135.1 | 4 | 1228 | |||
NM_001317207.1 | NP_001304136.1 | 2 | 910 | |||
NM_001317208.1 | NP_001304137.1 | 3 | 1016 | |||
NM_001317209.1 | NP_001304138.1 | 2 | 922 | |||
NM_001317210.1 | NP_001304139.1 | 3 | 1010 | |||
NM_001317211.1 | NP_001304140.1 | 3 | 1004 | |||
NM_016463.8 | NP_057547.5 | 2 | 1377 | |||
XM_017009573.1 | XP_016865062.1 | 2 | 1247 | |||
GenBank mRNA | AB097005.1 | - | 2 | 954 | ||
AB097032.1 | - | 2 | 954 | |||
AF151029.1 | - | 1 | 583 | |||
AK001782.1 | - | 2 | 899 | |||
AK024338.1 | - | 2 | 1377 | |||
AK314420.1 | - | 1 | 661 | |||
AY007103.1 | - | 1 | 1022 | |||
BC002490.2 | - | 1 | 713 | 3050465 | ||
BC006428.1 | - | 2 | 1308 | 3343683 | ||
BC017439.1 | - | 2 | 1269 | 2989328 | ||
BC024040.1 | - | 2 | 1298 | 3925666 | ||
BC041013.1 | - | 2 | 1298 | 4139961 | ||
BC050046.1 | - | 2 | 1270 | |||
GQ379202.1 | - | 1 | 663 |