Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003425.3 | NP_003416.1 | 10 | 1383 | ||
XM_011527267.1 | XP_011525569.1 | 11 | 1466 | |||
XM_011527269.1 | XP_011525571.1 | 10 | 1379 | |||
XM_011527271.1 | XP_011525573.1 | 10 | 1096 | |||
XM_011527273.1 | XP_011525575.1 | 9 | 1296 | |||
XM_017027217.1 | XP_016882706.1 | 12 | 1585 | |||
XM_017027218.1 | XP_016882707.1 | 12 | 1581 | |||
XM_017027219.1 | XP_016882708.1 | 11 | 1463 | |||
XM_017027220.1 | XP_016882709.1 | 12 | 1297 | |||
XM_017027221.1 | XP_016882710.1 | 12 | 1599 | |||
XM_017027222.1 | XP_016882711.1 | 11 | 1502 | |||
XM_017027223.1 | XP_016882712.1 | 12 | 1326 | |||
XM_017027224.1 | XP_016882713.1 | 10 | 1401 | |||
XM_017027225.1 | XP_016882714.1 | 11 | 1214 | |||
XM_017027226.1 | XP_016882715.1 | 11 | 1210 | |||
XM_017027227.1 | XP_016882716.1 | 9 | 1008 | |||
XM_017027228.1 | XP_016882717.1 | 10 | 1446 | |||
XM_017027229.1 | XP_016882718.1 | 11 | 1565 | |||
XM_017027230.1 | XP_016882719.1 | 10 | 1442 | |||
GenBank mRNA | AK127739.1 | - | 10 | 1383 | ||
BC037575.1 | - | 7 | 650 | 5539981 | ||
L75847.1 | - | 4 | 395 | |||
M67509.1 | - | 4 | 395 |