Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FGF4 SIRNA Assays ›
          Gene Symbol:
          Fgf4
          Gene Name:
          fibroblast growth factor 4
          Gene Aliases:
          Hst, Hst1
          Chromosome Location:
          Chr.1: 218025701 - 218027235 on Build Rnor_6.0
          UniGene ID:
          Rn.81223
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 46900

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Fgf4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_053809.1 NP_446261.1 2, 3 407
          XM_006230640.2 XP_006230702.1 2, 3 537
          GenBank mRNA AB079673.1 - 2, 3 407
          AF260830.1 - 1, 2 73

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_010202, AK082814, BC104312, BC104313, M30642.

          Gene Ontology Categories:

          Function(s) Process(es)

          cartilage condensation
          positive regulation of protein phosphorylation
          positive regulation of cell proliferation
          fibroblast growth factor receptor signaling pathway
          mesenchymal cell proliferation
          regulation of gene expression
          positive regulation of gene expression
          stem cell population maintenance
          embryonic limb morphogenesis
          embryonic hindlimb morphogenesis
          odontogenesis of dentin-containing tooth
          negative regulation of apoptotic process
          positive regulation of transcription from RNA polymerase II promoter
          cranial suture morphogenesis
          apoptotic process involved in morphogenesis
          chondroblast differentiation
          positive regulation of ERK1 and ERK2 cascade
          regulation of endothelial cell chemotaxis to fibroblast growth factor
          fibroblast growth factor receptor binding
          growth factor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0