Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapkapk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178102.2 | NP_835203.1 | 2 | 459 | ||
GenBank mRNA | AY197741.1 | - | 2 | 459 | ||
BC062048.1 | - | 2 | 425 | 5598069 |