Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012505.2 | NP_036637.1 | 12 | 1708 | ||
GenBank mRNA | BC085764.1 | - | 12 | 1708 | 7190910 | |
M14512.1 | - | 12 | 1693 |