Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053021.2 | NP_444180.2 | 9 | 1574 | ||
XM_006252094.2 | XP_006252156.1 | 8 | 1452 | |||
GenBank mRNA | AF314657.1 | - | 5 | 590 | ||
BC061534.1 | - | 8 | 1353 | 6886725 | ||
FQ209467.1 | - | 8 | 1353 | |||
FQ209498.1 | - | 8 | 1352 | |||
FQ209623.1 | - | 8 | 1349 | |||
FQ211071.1 | - | 8 | 1347 | |||
FQ213433.1 | - | 8 | 1356 | |||
FQ214098.1 | - | 8 | 1394 | |||
FQ218237.1 | - | 8 | 1350 | |||
FQ218812.1 | - | 8 | 1351 | |||
FQ219030.1 | - | 8 | 1348 | |||
M16975.1 | - | 9 | 1574 | |||
M64723.1 | - | 8 | 1339 | |||
X13231.1 | - | 8 | 1338 |