Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptger3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012704.1 | NP_036836.1 | 2 | 987 | ||
XM_006233535.2 | XP_006233597.1 | 2 | 1089 | |||
GenBank mRNA | AF302686.1 | - | 2 | 955 | ||
D14869.1 | - | 2 | 955 | |||
D16443.1 | - | 2 | 955 | |||
D29969.1 | - | 2 | 987 | |||
X80133.1 | - | 2 | 1004 | |||
X83855.1 | - | 2 | 1049 |
Set Membership: |
![]() |