Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CK357656.1 | - | 2 | 282 | ||
FQ217150.1 | - | 2 | 312 | |||
FQ217655.1 | - | 2 | 314 | |||
FQ220032.1 | - | 571 | ||||
FQ220162.1 | - | 2 | 313 | |||
FQ221038.1 | - | 2 | 311 | |||
FQ221154.1 | - | 2 | 302 | |||
FQ221373.1 | - | 2 | 312 | |||
FQ222161.1 | - | 2 | 312 | |||
FQ222277.1 | - | 2 | 225 | |||
FQ222316.1 | - | 2 | 311 | |||
FQ222356.1 | - | 2 | 312 | |||
FQ222897.1 | - | 2 | 313 | |||
FQ223053.1 | - | 2 | 311 | |||
FQ224290.1 | - | 2 | 313 | |||
FQ224782.1 | - | 2 | 312 | |||
FQ228448.1 | - | 2 | 311 | |||
FQ228589.1 | - | 2 | 312 | |||
M55534.1 | - | 851 | ||||
S74229.1 | - | 2 | 275 | |||
S77138.1 | - | 2 | 289 | |||
S77142.1 | - | 2 | 274 | |||
X60351.1 | - | 2 | 281 |