Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il1r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013123.3 | NP_037255.3 | 3 | 269 | ||
XM_006244746.2 | XP_006244808.1 | 4 | 2378 | |||
XM_006244747.2 | XP_006244809.1 | 4 | 635 | |||
XM_006244748.2 | XP_006244810.1 | 3 | 539 | |||
XM_006244749.2 | XP_006244811.1 | 5 | 458 | |||
XM_006244750.2 | XP_006244812.1 | 5 | 2450 | |||
XM_006244751.2 | XP_006244813.1 | 4 | 369 | |||
XM_006244752.2 | XP_006244814.1 | 4 | 611 | |||
XM_006244753.2 | XP_006244815.1 | 4 | 1730 | |||
XM_006244754.2 | XP_006244816.1 | 3 | 2282 | |||
XM_008766993.1 | XP_008765215.1 | 4 | 386 | |||
GenBank mRNA | AB112436.1 | - | 5 | 403 | ||
AB112437.1 | - | 3 | 269 | |||
AB112438.1 | - | 4 | 341 | |||
M95578.1 | - | 4 | 590 | |||
U14010.1 | - | 2 | 218 |