Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013218.1 | NP_037350.1 | 3 | 343 | ||
GenBank mRNA | BC064656.1 | - | 3 | 345 | 5621289 | |
D13062.1 | - | 3 | 343 |